Sodium Acetate 98% buffer solution
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/ reagent.
Synonyms: Acetic acid sodium salt
- CAS Number:126-96-5
- Formula:C₄H₇NaO₄
- Molecular Weight:142,09 g/mol
3 Options Available Below
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: Acetic acid sodium salt
- CAS Number:126-96-5
- Formula:C₄H₇NaO₄
- Molecular Weight:142,09 g/mol



