Orders LinkContactUsLinkComponent
NBD-556 98%
New Product
NBD-556 98%
New Product
NBD-556 98%
Usage: Organic building block/ reagent.
 :  N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethylpiperidin-4-yl)oxalamide, N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)ethanediamide
  •  :333353-44-9
  •  :C₁₇H₂₄ClN₃O₂
  •  :337,84 g/mol
Product Family Options
  • Pack type
  • Bottle
    1 * 100 mg
    1 * 100 mg - 216,00 €
    216,00 €
    1 * 100 mg
       216,00 €
    New Product
    New Product
  • Bottle
    1 * 250 mg
    1 * 250 mg - 472,00 €
    472,00 €
    1 * 250 mg
       472,00 €
    New Product
    New Product
  • Bottle
    1 * 1 g
    1 * 1 g - 1 280,00 €
    1 280,00 €
    1 * 1 g
       1 280,00 €