Fluorescein indicator
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/ reagent.
Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
- CAS Number:2321-07-5
- Formula:C₂₀H₁₂O₅
- MDL Number:MFCD00005050
- Molecular Weight:332,31 g/mol
- EINECS:219-031-8
4 Options Available Below
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
- CAS Number:2321-07-5
- Formula:C₂₀H₁₂O₅
- MDL Number:MFCD00005050
- Molecular Weight:332,31 g/mol
- EINECS:219-031-8
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



