Order Entry
Export
Orders LinkContactUsLinkComponent
Vinylferrocene 98%

Vinylferrocene 98%

Flammable
Supplier:   Apollo Scientific
Vinylferrocene 98%
Supplier:  Apollo Scientific
Certificates
Vinylferrocene 98%

6 Options Available Below

Product Details & Documents

Usage: Inorganic salt or catalyst.

  • Store at 2 to 8 °C
  • CAS Number:1271-51-8
  • Formula:C₁₂H₁₂Fe
  • MDL Number:MFCD00001434
  • Molecular Weight:212,07 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:215-041-1
  • UN:1325
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 24,60 €
    24,60 €
    1 * 1 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 75,30 €
    75,30 €
    1 * 5 g
       75,30 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 343,00 €
    343,00 €
    1 * 25 g
       343,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1 260,00 €
    1 260,00 €
    1 * 100 g
       1 260,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 3 530,00 €
    3 530,00 €
    1 * 500 g
       3 530,00 €

    Specifications:
    New Product
    New Product