Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
1,12-Dodecanedioic acid
BP0000689751
Dodecanedioic acid
Print
Dodecanedioic acid
New Product
Supplier:
Apollo Scientific
Dodecanedioic acid
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
Dodecanedioic acid
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
2 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP19772-5G
Bottle
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 69,50 €
69,50 €
-
+
1 * 5 g
69,50 €
-
+
Specifications:
New Product
New Product
APOSBUP19772-10G
Bottle
Log in
for availability
Log in
for availability
1 * 10 g
1 * 10 g - 102,00 €
102,00 €
-
+
1 * 10 g
102,00 €
-
+
Specifications:
New Product
New Product