4-Hydroxyindole ≥95%
Bottle
Catalog # APOSBUP18962-1G
New Product
Supplier: Apollo Scientific
4-Hydroxyindole ≥95%
Catalog # APOSBUP18962-1G
Supplier: Apollo Scientific
New Product
CAS Number: 2380-94-1
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol
- CAS Number:2380-94-1
- Formula:C₈H₇NO
- Molecular Weight:133,15 g/mol
Specifications
Specifications
- Pack type:Bottle



