Order Entry
Export
Orders LinkContactUsLinkComponent
Bathophenanthroline ≥98%

Bathophenanthroline ≥98%

Bottle
Catalog # APOSBUP18829-500MG
New Product
Supplier:   Apollo Scientific
Bathophenanthroline ≥98%
Catalog # APOSBUP18829-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1662-01-7
Bathophenanthroline ≥98%

Bottle

CAS Number:  1662-01-7
Price
37,70 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)
  • CAS Number:1662-01-7
  • Formula:C₂₄H₁₆N₂
  • Molecular Weight:332,4 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications