Order Entry
Export
Orders LinkContactUsLinkComponent
8-Hydroxyquinoline ≥98%

8-Hydroxyquinoline ≥98%

New Product
Supplier:   Apollo Scientific
8-Hydroxyquinoline ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
8-Hydroxyquinoline ≥98%

4 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  8-Hydroxyquinoline
  • CAS Number:148-24-3
  • Formula:C₉H₇NO
  • Molecular Weight:145,16 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 40,60 €
    40,60 €
    1 * 500 mg
       40,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 49,20 €
    49,20 €
    1 * 1 g
       49,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 62,30 €
    62,30 €
    1 * 5 g
       62,30 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 86,90 €
    86,90 €
    1 * 10 g
       86,90 €

    Specifications:
    New Product
    New Product