Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
3-Hydroxycinnamic acid
BP0000689604
m-Coumaric acid
Print
m-Coumaric acid
New Product
Supplier:
Apollo Scientific
m-Coumaric acid
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
m-Coumaric acid
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
2 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP18246-500MG
Bottle
Log in
for availability
Log in
for availability
1 * 500 mg
1 * 500 mg - 63,70 €
63,70 €
-
+
1 * 500 mg
63,70 €
-
+
Specifications:
New Product
New Product
APOSBUP18246-10G
Bottle
Log in
for availability
Log in
for availability
1 * 10 g
1 * 10 g - 280,00 €
280,00 €
-
+
1 * 10 g
280,00 €
-
+
Specifications:
New Product
New Product