Squalene ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/ life science applications.
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
- CAS Number:111-02-4
- Formula:C₃₀H₅₀
- Molecular Weight:410,72 g/mol
2 Options Available Below
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
- CAS Number:111-02-4
- Formula:C₃₀H₅₀
- Molecular Weight:410,72 g/mol



