Order Entry
Export
Orders LinkContactUsLinkComponent
 

Protocatechualdehyde ≥98%

New Product
Supplier:   Apollo Scientific
Protocatechualdehyde ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Protocatechualdehyde ≥98%

5 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Protocatechuic aldehyde, Protocatechualdehyde
  • CAS Number:139-85-5
  • Formula:C₇H₆O₃
  • Molecular Weight:138,12 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 52,10 €
    52,10 €
    1 * 100 mg
       52,10 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 118,00 €
    118,00 €
    1 * 500 mg
       118,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 170,00 €
    170,00 €
    1 * 1 g
       170,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 380,00 €
    380,00 €
    1 * 5 g
       380,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 563,00 €
    563,00 €
    1 * 10 g
       563,00 €

    Specifications:
    New Product
    New Product