Order Entry
Export
Orders LinkContactUsLinkComponent
L-Thyroxine 97%

L-Thyroxine 97%

Health
New Product
Supplier:   Apollo Scientific
L-Thyroxine 97%
Supplier:  Apollo Scientific
New Product
Certificates

6 Options Available Below

Product Details & Documents

Usage: Organic building block/ reagent.

  • Store at 2 to 8 °C
Synonyms:  (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
  • CAS Number:51-48-9
  • Formula:C₁₅H₁₁I₄NO₄
  • MDL Number:MFCD00002595
  • Molecular Weight:776,87 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:200-101-1
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 24,60 €
    24,60 €
    1 * 1 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 49,20 €
    49,20 €
    1 * 5 g
       49,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 232,00 €
    232,00 €
    1 * 25 g
       232,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 879,00 €
    879,00 €
    1 * 100 g
       879,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 3 600,00 €
    3 600,00 €
    1 * 500 g
       3 600,00 €

    Specifications:
    New Product
    New Product