Order Entry
Export
Orders LinkContactUsLinkComponent
 

Alvespimycin ≥98%

New Product
Supplier:   Apollo Scientific
Alvespimycin ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Alvespimycin ≥98%

3 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  17-Dimethylaminoethylamino-17-demethoxygeldanamycin
  • CAS Number:467214-20-6
  • Formula:C₃₂H₄₈N₄O₈
  • Molecular Weight:616,75 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 368,00 €
    368,00 €
    1 * 25 mg
       368,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 545,00 €
    545,00 €
    1 * 50 mg
       545,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 793,00 €
    793,00 €
    1 * 100 mg
       793,00 €

    Specifications:
    New Product
    New Product