Order Entry
Export
Orders LinkContactUsLinkComponent
Antitubercular agent-30 ≥98%

Antitubercular agent-30 ≥98%

New Product
Supplier:   Apollo Scientific
Antitubercular agent-30 ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Antitubercular agent-30 ≥98%

4 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Antitubercular agent-30
  • CAS Number:384857-54-9
  • Formula:C₁₂H₁₀N₂O₃S
  • Molecular Weight:262,28 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 202,00 €
    202,00 €
    1 * 25 mg
       202,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 301,00 €
    301,00 €
    1 * 50 mg
       301,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 440,00 €
    440,00 €
    1 * 100 mg
       440,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 620,00 €
    620,00 €
    1 * 200 mg
       620,00 €

    Specifications:
    New Product
    New Product