Order Entry
Export
Orders LinkContactUsLinkComponent
BIOFLOAT™ 384-Well Plates

BIOFLOAT™ 384-Well Plates

BIOFLOAT™ 384-well plate, polystyrene, 16×24 array, 384, Clear, 90 μl, +, Individually packed
Catalog # FCELF221384-1
New Product
Supplier:   FACELLITATE
BIOFLOAT™ 384-Well Plates
Catalog # FCELF221384-1
Supplier:  FACELLITATE
New Product
BIOFLOAT™ 384-Well Plates

BIOFLOAT™ 384-well plate, polystyrene, 16×24 array, 384, Clear, 90 μl, +, Individually packed

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

The BIOFLOAT™ 384-well plate provides a highly defined, fully inert, and cell-repelling surface.

The coating modifies the plastic surface to cell and protein repellent surface to facilitate rapid spheroid and organoid formation. The plates were sterilized using electron beam irradiation according to DIN EN ISO 11137.

The BIOFLOAT™ 384-well plate outperforms existing products by its anti-adhesive properties:

 

Rapid spheroid formation – BIOFLOAT plates are supporting faster cell aggregation into spheroids compared to competitor plates. Spheroids are usually forming within 24 hours after cell seeding in BIOFLOAT plates.

High reproducibility of spheroid generation – A single perfectly round-shaped spheroid is forming in each well of BIOFLOAT plates, without any further satellites or cell aggregates.

 

Reliable spheroid formation – Spheroids can be generated in BIOFLOAT plates with a variety of different cell types, including tumor cell lines, human and animal primary cells and stem cells.

No need for centrifugation – Centrifugation after cell seeding is not needed, saving time and reducing cellular stress.

 

Stability in long term cultivation of spheroids – Highly viable and functional spheroids can be cultivated for several months in BIOFLOAT plates.

Caution: For Research use only. Not intended for diagnostic or therapeutic use.

Specifications
  • No. of wells:
    384
  • Packed:
    Individually packed
  • Sterile:
    +
  • Colour:
    Clear
  • Description:
    BIOFLOAT™ 384-well plate, polystyrene, 16×24 array
  • Well volume:
    90 μl
  • Cat. No.:
    FCELF221384-1
  • Storage temperature:
    −20....+50 °C
  • Dimensions:
    85,48×W127,67H×14,4L mm