0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
STF-31
BP0000683655
STF-31
New Product
:
Apollo Scientific
STF-31
:
Apollo Scientific
New Product
STF-31
Product Family Options
Pack type
APOSBUP07777-25MG
Bottle
1 * 25 mg
1 * 25 mg - 310,00 €
310,00 €
-
+
1 * 25 mg
310,00 €
-
+
New Product
New Product
APOSBUP07777-50MG
Bottle
1 * 50 mg
1 * 50 mg - 585,00 €
585,00 €
-
+
1 * 50 mg
585,00 €
-
+
New Product
New Product
APOSBUP07777-100MG
Bottle
1 * 100 mg
1 * 100 mg - 856,00 €
856,00 €
-
+
1 * 100 mg
856,00 €
-
+
New Product
New Product