Order Entry
Export
Orders LinkContactUsLinkComponent
DL-alpha-Tocopherol 97%

DL-alpha-Tocopherol 97%

Irritant
Supplier:   Apollo Scientific
DL-alpha-Tocopherol 97%
Supplier:  Apollo Scientific
Certificates
DL-alpha-Tocopherol 97%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

  • Store at −20 °C
Synonyms:  (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E
  • CAS Number:10191-41-0
  • Formula:C₂₉H₅₀O₂
  • MDL Number:MFCD00072051
  • Molecular Weight:430,70609 g/mol
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 29,00 €
    29,00 €
    1 * 25 g
       29,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 94,10 €
    94,10 €
    1 * 100 g
       94,10 €