Dilute Bismuth Subnitrate-Potassium Iodide, Test Solution (TS)
Catalog # ACSDJPH223
New Product
Supplier: CPACHEM
Dilute Bismuth Subnitrate-Potassium Iodide, Test Solution (TS)
Catalog # ACSDJPH223
Supplier: CPACHEM
New Product
CAS Number: 87-69-4
Jump to:
Product Details & Documents
Dilute bismuth subnitrate-potassium iodide TS for spraying
Shelf Life on Ship Date: 12 Months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.
- CAS Number:87-69-4
- EINECS:201-766-0



