Order Entry
Export
Orders LinkContactUsLinkComponent
Palletank® Drum for Storage

Palletank® Drum for Storage

Drum with 1 bottom gate, 200 L, HDPE, 918 mm (36,1")
Catalog # SARTFBC125629
New Product
Supplier:   Sartorius
Palletank® Drum for Storage
Catalog # SARTFBC125629
Supplier:  Sartorius
New Product
Palletank® Drum for Storage

Drum with 1 bottom gate, 200 L, HDPE, 918 mm (36,1")

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Palletank® for small volume storage are stainless steel containers designed for the reliable and robust storage of biopharmaceutical fluids such as high value drug substances contained in Flexsafe® 3D Bags. The Flexsafe® 3D Bags are manufactured according to a qualified design that precisely fits the Palletank®.

  • Applications: 

    Liquid storage and shipping

    Media preparation

    Buffer preparation

    Drug substance purification

    Drug product formulation

    Cell harvest and downstream processing intermediates

The double-hinged front doors allow for bag replacement, installation, and manipulation while Palletank® are stacked up in the process area, thus enabling continuous processing without movement of the containers. As a result, a forklift is not routinely required in the manufacturing area.

Specifications
  • Ext.Ø:
    918 mm (36,1")
  • Capacity:
    200 L
  • Description:
    Drum with 1 bottom gate
  • Material:
    HDPE
  • Seq:
    2
  • Cat. No.:
    SARTFBC125629
  • Int.Ø:
    559 mm (22")