BIO-LAB-TOP
Supplier: MACHEREY-NAGEL
Jump to:
About this item
Protect laboratory surfaces from spills.
BIO-LAB-TOP is a filter paper used for surface protection from spills
- The filter paper is coated on one side with polyethylene
The filter paper has a water adsorption of 210 to 230 ml/m².
Shelf life: 72 months from production date.
5 Options Available Below
Product Details & Documents
Protect laboratory surfaces from spills.
BIO-LAB-TOP is a filter paper used for surface protection from spills
- The filter paper is coated on one side with polyethylene
The filter paper has a water adsorption of 210 to 230 ml/m².
Shelf life: 72 months from production date.
Product Family Options
Sheets
Product Information
- L×WThicknessAbsorbencyAvailabilityPrice
- MANA740800Filter paper, 140 g/m², one side coated with polyethyleneFilter paper, 140 g/m², one side coated with polyethylene480×600 mm0.22 mm/8.7 mil210 - 230 ml/m²Log in for availability1 * 50 items102,00 €
- MANA740801Filter paper, 140 g/m², one side coated with polyethyleneFilter paper, 140 g/m², one side coated with polyethylene480×600 mm0.22 mm/8.7 mil210 - 230 ml/m²Log in for availability1 * 100 items174,00 €
Rolls
Product Information
- L×WThicknessAbsorbencyAvailabilityPrice
- MANA740810Filter paper, 140 g/m², one side coated with polyethyleneFilter paper, 140 g/m², one side coated with polyethylene0.48×50 m0.22 mm/8.7 mil210 - 230 ml/m²Log in for availability1 * 1 Roll151,00 €
- MANA740820Filter paper, 140 g/m², one side coated with polyethyleneFilter paper, 140 g/m², one side coated with polyethylene0.6×50 m0.22 mm/8.7 mil210 - 230 ml/m²Log in for availability1 * 1 Roll178,00 €
- MANA740821Filter paper, 140 g/m², one side coated with polyethyleneFilter paper, 140 g/m², one side coated with polyethylene0.6×100 m0.22 mm/8.7 mil210 - 230 ml/m²Log in for availability1 * 1 Roll294,00 €