Order Entry
Export
Orders LinkContactUsLinkComponent
Acid Wash/Regenerating Solution

Acid Wash/Regenerating Solution

Acid wash regenerating solution, 100 ml
Catalog # EKSI5312349
New Product
Supplier:   EKSIGENT
Acid Wash/Regenerating Solution
Catalog # EKSI5312349
Supplier:  EKSIGENT
New Product
Acid Wash/Regenerating Solution

Acid wash regenerating solution, 100 ml

Price
Inquire For Price
Log in for availability
1 * 100 mL
Product Details & Documents

Standalone acid (100 ml 0,1 N HCl).

his acid is also sold in the following kits: P/N 390953, SDS-MW kit P/N A10663, IgG purity and heterogeneity kit P/N C44790, CZE rapid charge variant analysis kit P/N C48231, RNA 9000 purity and integrity kit P/N C30085, BioPhase CE-SDS protein analysis kit.

Specifications
  • Description:
    Acid wash regenerating solution, 100 ml