Orders LinkContactUsLinkComponent
Micro Powder Spatulas, Angular/Semi-Cup-Shaped
Micro Powder Spatulas, Angular/Semi-Cup-Shaped
New Product
Supplier:   BURKLE
Micro Powder Spatulas, Angular/Semi-Cup-Shaped
Supplier:  BURKLE
New Product
Certificates
Micro Powder Spatulas, Angular/Semi-Cup-Shaped
2 Options Available Below
Product Details & Documents
The micro powder spatula is made of corrosion-resistant stainless steel 1.4301 (304) and is specially designed for precise dosing and contamination-free work in analytical laboratories. Its slim shape and dual-sided design allow for precise handling of small powder quantities, even in narrow containers such as vials or test tubes.
  • Two functions: flat side for spreading, semicircular side for picking up and transporting
    Slim shaft enables work in narrow vessels such as vials, test tubes and bottles
    Fine blade for precise dosing and pinpoint accuracy when working with very small quantities
The polished surface prevents residue buildup and makes cleaning easy. This spatula is an indispensable tool for applications requiring precision and hygiene, such as sample preparation, weighing sensitive substances, and spreading on analysis surfaces.
The micro powder spatula is ideal for use in chemical, pharmaceutical, and analytical laboratories, where precise and safe processing of small quantities of powder is necessary. Typical applications include preparing samples for spectroscopic analysis, weighing sensitive substances, and working in narrow vessels. The spatula is also used in environmental, food, and material analysis.
Product Family Options
Product Information
  • Length
    Colour
    Availability
    Price
  • BURK5386-0096
    Micro powder spatula, angular/semi-cup-shaped
    Micro powder spatula, angular/semi-cup-shaped
    150 mm
    Silver
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1,90 €
    1,90 €
    1 * 1 items
       1,90 €
    Specifications:
    New Product
    New Product
  • BURK5386-0098
    Micro powder spatula, angular/semi-cup-shaped
    Micro powder spatula, angular/semi-cup-shaped
    210 mm
    Silver
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 2,30 €
    2,30 €
    1 * 1 items
       2,30 €
    Specifications:
    New Product
    New Product