Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl cellulose FCC

Ethyl cellulose FCC

New Product
Supplier:   Spectrum Chemical
Ethyl cellulose FCC
Supplier:  Spectrum Chemical
New Product
Certificates

3 Options Available Below

Product Details & Documents

ETHYL CELLULOSE, FCC, a derivative of cellulose, is often used as a known as an emulsifier food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

  • Storage temperature - Keep tightly closed
Synonyms:  Ethylcellulose
  • CAS Number:9004-57-3
  • MDL Number:MFCD00131037
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Assay (Ethoxy Groups; Dried Basis):
    44,0 - 50,0%
  • Viscosity:
    To Pass Test
  • Lead (Pb):
    max. 3 mg/kg
  • Loss On Drying:
    max. 3%
  • Residue On Ignition:
    max. 0,5%

Specifications

Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Poly bottle
    Log in for availability
    Log in for availability
    1 * 125 g
    1 * 125 g

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    SPCME1036-125G
    New Product
  • Poly bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    SPCME1036-500G
    New Product
  • Poly pail
    Log in for availability
    Log in for availability
    1 * 2.500 g
    1 * 2.500 g

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    SPCME1036-2.5KG
    New Product