Orders LinkContactUsLinkComponent
External Dosing Station
Equipment constructed and designed for use in conjunction with media preparators inside a laminar flow hood in laboratories that dispense culture media DW-MP-TS, for automation and acceleration of the dispensing phase in repetitive operations of medium and large volumes; 210×285×200 mm (L×D×H); 2,85 kg; 50 W heating capacity; 90 - 250 V, 50/60 Hz
  ESPIDW-MP-TS
New Product
 :   RAYPA
External Dosing Station
  ESPIDW-MP-TS
 :  RAYPA
New Product
External Dosing Station
External Dosing Station
Equipment constructed and designed for use in conjunction with media preparators inside a laminar flow hood in laboratories that dispense culture media DW-MP-TS, for automation and acceleration of the dispensing phase in repetitive operations of medium and large volumes; 210×285×200 mm (L×D×H); 2,85 kg; 50 W heating capacity; 90 - 250 V, 50/60 Hz
1 * 1 items
Equipment constructed and designed for use in conjunction with media preparators inside a laminar flow hood in laboratories that dispense culture media.
  • Included items: control box with dosage selection, dosing pedal, optical sensor, power cable, sensor connection cable and instruction manual
  • Modern touchscreen display that’s user-friendly and easy to navigate
  • Automatic self-cleaning and disinfection of the dispensing lines
  • Possibility of transferring the dispensing line through barriers such as glass or walls, making it ideal for use in laminar flow hoods or clean rooms
  • This system allows for significantly higher dispensing speeds, ranging between 65 to 100 ml per second
Compatible with dispensing lines of Ø 6,4, 4,8 and 4 mm, there are two mechanisms available for triggering each dispensation: either manually using a foot pedal or through a semi-automatic process activated by an optical sensor.
  • Description:
    Equipment constructed and designed for use in conjunction with media preparators inside a laminar flow hood in laboratories that dispense culture media DW-MP-TS, for automation and acceleration of the dispensing phase in repetitive operations of medium and large volumes; 210×285×200 mm (L×D×H); 2,85 kg; 50 W heating capacity; 90 - 250 V, 50/60 Hz