Order Entry
Export
Orders LinkContactUsLinkComponent
PBFI AM ≥94% fluorescent potassium dye

PBFI AM ≥94% fluorescent potassium dye

Catalog # ABCAAB142804-1
Supplier:   Abcam
PBFI AM ≥94% fluorescent potassium dye
Catalog # ABCAAB142804-1
Supplier:  Abcam
CAS Number:  124549-23-1
PBFI AM ≥94% fluorescent potassium dye
CAS Number:  124549-23-1
Price
549,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Cell-permeable, fluorescent K+ binding probe (Kd = 5 mM). Active *in vitro*. Non-specific esterases hydrolyze the AM esters to produce the water-soluble form of PBFI.Excitation is 340/390 nm.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Pbfi-AM
  • CAS Number:124549-23-1
  • Molecular Weight:1171,1 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% PBFI AM