Orders LinkContactUsLinkComponent
Premium Casters
Stronger, medical grade casters that include brakes, 4WHBR, Enhancing the mobility of the equipment
Catalog # ESPI4WHBR
Supplier:   RAYPA
Premium Casters
Catalog # ESPI4WHBR
Supplier:  RAYPA
Premium Casters
Premium Casters
Stronger, medical grade casters that include brakes, 4WHBR, Enhancing the mobility of the equipment
Price
161,00 €
Log in for availability
1 * 4 items
Product Details & Documents
Stronger, medical grade casters that include brakes.
  • This enhances the mobility of the equipment.
Although all AE-DRY and AES autoclaves include casters, this accessory offers the option of upgrading to stronger, medical-grade casters that include brakes.
 : Factory installation required.
Specifications
  • Description:
    Stronger, medical grade casters that include brakes, 4WHBR
  • For:
    Enhancing the mobility of the equipment