Order Entry
Export
Orders LinkContactUsLinkComponent
LM11A-31 nonpeptide p75NTR ligand

LM11A-31 nonpeptide p75NTR ligand

Supplier:   Abcam
LM11A-31 nonpeptide p75NTR ligand
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

Selective nonpeptide p75NTR ligand. Blocks p75-mediated cell death. Promotes neurogenesis and neurite survival. Does not affect NGF binding to TrkA. Shows neuroprotective and neuroregenerative effects in vivo. Orally active. Blood-brain barrier permeable.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (2S,3S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide, dihydrochloride
  • CAS Number:1243259-19-9
  • Molecular Weight:316,26 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% LM11A-31
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 375,00 €
    375,00 €
    1 * 25 mg
       375,00 €

  • Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 103,00 €
    103,00 €
    1 * 5 mg
       103,00 €