Orders LinkContactUsLinkComponent
Metergoline ≥99% 5-HT receptor antagonist
 :   Abcam
Metergoline ≥99% 5-HT receptor antagonist
 :  Abcam
Metergoline ≥99% 5-HT receptor antagonist
Potent 5-HT₁, 5-HT₂ and 5-HT₇ receptor antagonist (Ki = 16 nM at h5-HT₇ receptor). Additionally displays activity at dopamine receptors. Elicits cell death.
 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
 :  [(8β)-1,6-Dimethylergolin-8-yl)methyl]carbamic acid phenylmethyl ester
  •  :17692-51-2
  •  :403,5 g/mol
  • Constituents::
    100% Metergoline
Product Family Options
  •  
  •    
    1 * 10 mg
    1 * 10 mg - 66,70 €
    66,70 €
    1 * 10 mg
       66,70 €
  •    
    1 * 50 mg
    1 * 50 mg - 241,00 €
    241,00 €
    1 * 50 mg
       241,00 €