Order Entry
Export
Orders LinkContactUsLinkComponent
Dynole® 31-2 ≥97% Negative control for Dynolereg 34-2

Dynole® 31-2 ≥97% Negative control for Dynolereg 34-2

Catalog # ABCAAB120464-5
Supplier:   Abcam
Dynole® 31-2 ≥97% Negative control for Dynolereg 34-2
Catalog # ABCAAB120464-5
Supplier:  Abcam
CAS Number:  
Dynole® 31-2 ≥97% Negative control for Dynolereg 34-2
CAS Number:  
Price
231,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

Negative control molecule for use with Dynole® 34-2. Displays no significant activity at dynamin I or II (up to 300 µM).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • Molecular Weight:338,45 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Dynole-31-2™