MALDI Biotyper® Biotargets
Individually barcoded disposable 96 position MALDI target plates, Box of 20
Supplier: BRUKER DALTRONIK
Jump to:
Product Details & Documents
The MBT Biotargets are equipped with hydrophobic coating on the target surrounding the hydrophilic sample positions facilitates the preparation of samples.
Disposable MALDI target plates
96 sample positions
Unique barcode for traceability
Perfectly flat surface for reliable and reproducible results
High flexibility in sample run sizes
No wasted spots
AnchorChip™ effect facilitates preparation
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:Individually barcoded disposable 96 position MALDI target plates
- Pk Info:Box of 20