Hygromycin B
Supplier: G-Biosciences
Jump to:
About this item
Mode of action and Anti-microbial spectrum: Hygromycin B is an aminocyclitol with a wide spectrum of activity that kills bacteria, fungi and higher eukaryotic cells by inhibiting protein synthesis. It is produced by the bacterium Streptomyces hygroscopicus.
- Molecular Biology Grade
- Tested against both sensitive and resistant cells
Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
- CAS Number:31282-04-9
- Formula:C₂₀H₃₇N₃O₁₃
- Molecular Weight:527,6 g/mol
6 Options Available Below
Product Details & Documents
Mode of action and Anti-microbial spectrum: Hygromycin B is an aminocyclitol with a wide spectrum of activity that kills bacteria, fungi and higher eukaryotic cells by inhibiting protein synthesis. It is produced by the bacterium Streptomyces hygroscopicus.
- Molecular Biology Grade
- Tested against both sensitive and resistant cells
Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
- CAS Number:31282-04-9
- Formula:C₂₀H₃₇N₃O₁₃
- Molecular Weight:527,6 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice



