Cable Glands
New Product
Supplier: RAYPA
Jump to:
About this item
Installation of a Ø2 mm or Ø4 mm cable gland to provide access to as many as eight external temperature probes for calibration and validation procedures.
- Flexible and simple installation at any time, adapting to the user's needs without complications
- Compatible with Classic line and Top line autoclaves
- Two available options: GC2MM (up to 8 × Ø2 mm probes) and GC4MM (up to 8 × Ø4 mm probes)
2 Options Available Below
Product Details & Documents
Installation of a Ø2 mm or Ø4 mm cable gland to provide access to as many as eight external temperature probes for calibration and validation procedures.
- Flexible and simple installation at any time, adapting to the user's needs without complications
- Compatible with Classic line and Top line autoclaves
- Two available options: GC2MM (up to 8 × Ø2 mm probes) and GC4MM (up to 8 × Ø4 mm probes)
Product Family Options
Product Information
- ForAvailabilityPrice
- ESPICG4MMSpecial adaptation with the installation of a Ø 4 mm cable gland, allowing the insertion of up to eight external temperature probes CG4MMSpecial adaptation with the installation of a Ø 4 mm cable gland, allowing the insertion of up to eight external temperature probes CG4MMCalibration and validation proceduresLog in for availability1 * 1 itemsSpecifications:New ProductNew Product
- ESPICG2MMSpecial adaptation with the installation of a Ø 2 mm cable gland, allowing the insertion of up to eight external temperature probes CG2MMSpecial adaptation with the installation of a Ø 2 mm cable gland, allowing the insertion of up to eight external temperature probes CG2MMCalibration and validation proceduresLog in for availability1 * 1 itemsSpecifications:New ProductNew Product