Explorer™ Semi-Micro Balances
This new generation of laboratory balances is designed to enhance your workflow with unparalleled user experience, outstanding weighing performance, advanced communication capabilities, and an eco-friendly design.
7" touchscreen display; vibrant colours, an easy-to-clean glass panel, and chemical resistance ensure a clear and durable interface
- Effortless setup and monitoring; with a digital-level bubble, that ensures a smooth and straightforward setup
- Intuitive sample handling and reliable status indicators; features an intuitive sample spotlight that provides clear visibility, ensuring precise and hassle-free sample handling
- Customised terminal and lights; provide clear and immediate feedback to underload or overload conditions
- Internal Calibration Systems; Automatic, user-triggered, and span calibration options ensure precise measurements
Engineered for accuracy, designed for demanding applications: Standard Square Weighing Pan - designed for laboratory efficiency and specifically designed to assist in sample preparation. Sample Libraries shared among applications: Apply to 11 weighing applications and 1000 sample libraries for Explorer EXR.
Removable Draft shield: For easy cleaning and long-term care and specific large volume weighing applications. Digital High-Speed Load Cell; The digital weighing components (DWC) offer the advantage of creating self-contained load cell modules. Rapid and precise measurements with this advanced technology ensures weighing processes are both swift and highly accurate.
Anti-Static Draft shield: Reduces static electricity around the balance for consistent results. Routine Repeatability Test: Ensures consistent long-term accuracy and reliability. Ideal for critical applications such as weighing in regulated environments, where meeting strict uncertainty requirements is essential (e.g., pharmacopeia mandates uncertainty ≤ 0,10%).
Supports compliance for data traceability: Equipped with multiple USB ports; offering unparalleled flexibility, the direct to Excel feature provides a basic yet highly user-friendly solution for data transfer. Human Interface Device allows connections to computers without the need for drivers and easy integration with LIMS and MES systems. Time Synchronisation - Network Time Protocol (NTP) ensures accurate timekeeping. System Log: All the changes made on balance setting will keep records in system log file. These system log files can be reviewed and exported as a PDF to a USB flash drive. Direct Data Transfer; Direct export to Excel and saving data to USB in CSV and PDF formats. Label Printing Function: 2 preset templates and 10 customisable templates for semi-finished product samples transfer management. When the Electronic Signature feature is enabled, the User ID will be used in the printout field when the User ID content option is selected.
Eco-Friendly Design: 65% reduced power consumption in Sleep Mode. Reduced shipping weight; 36% weight reduction for non-draft shield models, 64% weight reduction for draft shield models. Recycled materials, up to 58% of total components. Aluminum construction: Replaces most plastic materials for durability and sustainability. Dematerialisation: Reduces the number of components to improve production efficiency. ACT Label Certification: Eleven models are certified with the ACT Label, highlighting their environmental impact.
: OIML R76, EN 45501 IEC/EN 61010-1; CAN/CSA C22.2 61010-1 UL 61010-1 IEC/EN 61326-1 Class B, Basic Environments FCC Part 15 Class A Canada ICES-003 Class A
: Supplied with AC adapter and a plastic in use protective cover.
This new generation of laboratory balances is designed to enhance your workflow with unparalleled user experience, outstanding weighing performance, advanced communication capabilities, and an eco-friendly design.
7" touchscreen display; vibrant colours, an easy-to-clean glass panel, and chemical resistance ensure a clear and durable interface
- Effortless setup and monitoring; with a digital-level bubble, that ensures a smooth and straightforward setup
- Intuitive sample handling and reliable status indicators; features an intuitive sample spotlight that provides clear visibility, ensuring precise and hassle-free sample handling
- Customised terminal and lights; provide clear and immediate feedback to underload or overload conditions
- Internal Calibration Systems; Automatic, user-triggered, and span calibration options ensure precise measurements
Engineered for accuracy, designed for demanding applications: Standard Square Weighing Pan - designed for laboratory efficiency and specifically designed to assist in sample preparation. Sample Libraries shared among applications: Apply to 11 weighing applications and 1000 sample libraries for Explorer EXR.
Removable Draft shield: For easy cleaning and long-term care and specific large volume weighing applications. Digital High-Speed Load Cell; The digital weighing components (DWC) offer the advantage of creating self-contained load cell modules. Rapid and precise measurements with this advanced technology ensures weighing processes are both swift and highly accurate.
Anti-Static Draft shield: Reduces static electricity around the balance for consistent results. Routine Repeatability Test: Ensures consistent long-term accuracy and reliability. Ideal for critical applications such as weighing in regulated environments, where meeting strict uncertainty requirements is essential (e.g., pharmacopeia mandates uncertainty ≤ 0,10%).
Supports compliance for data traceability: Equipped with multiple USB ports; offering unparalleled flexibility, the direct to Excel feature provides a basic yet highly user-friendly solution for data transfer. Human Interface Device allows connections to computers without the need for drivers and easy integration with LIMS and MES systems. Time Synchronisation - Network Time Protocol (NTP) ensures accurate timekeeping. System Log: All the changes made on balance setting will keep records in system log file. These system log files can be reviewed and exported as a PDF to a USB flash drive. Direct Data Transfer; Direct export to Excel and saving data to USB in CSV and PDF formats. Label Printing Function: 2 preset templates and 10 customisable templates for semi-finished product samples transfer management. When the Electronic Signature feature is enabled, the User ID will be used in the printout field when the User ID content option is selected.
Eco-Friendly Design: 65% reduced power consumption in Sleep Mode. Reduced shipping weight; 36% weight reduction for non-draft shield models, 64% weight reduction for draft shield models. Recycled materials, up to 58% of total components. Aluminum construction: Replaces most plastic materials for durability and sustainability. Dematerialisation: Reduces the number of components to improve production efficiency. ACT Label Certification: Eleven models are certified with the ACT Label, highlighting their environmental impact.
: OIML R76, EN 45501 IEC/EN 61010-1; CAN/CSA C22.2 61010-1 UL 61010-1 IEC/EN 61326-1 Class B, Basic Environments FCC Part 15 Class A Canada ICES-003 Class A
: Supplied with AC adapter and a plastic in use protective cover.
- Catalog No:
- OHAU31059121
- OHAU31059122
- OHAU31059123
- OHAU31059124
- OHAU31059132
- OHAU31059133
- OHAU31059134
- OHAU31059135
- Model:
- EXR125D
- EXR125
- EXR225D
- EXR225
- EXR125DM
- EXR125M
- EXR225DM
- EXR225M
- Operating temperature range:
- 10...30 °C
- 10...30 °C
- 10...30 °C
- 10...30 °C
- 10...30 °C
- 10...30 °C
- 10...30 °C
- 10...30 °C
- Communication interface:
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Voltage:
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
Product Family Options
- ModelWeighing cap.ReadabilityPan/Platform SizeRepeatabilityLinearityStabilisation timeDimensions
- EXR125D82 /120 g0,01 mg / 0,1 mg80×80 mm0,1 mg0,2 mg5 s372×408×229 mm1 * 1 items6 090,00 €New Product
Specifications
Cat. No.OHAU31059121ModelEXR125DOperating temperature range10...30 °CCommunication interfaceEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Voltage100 - 240 VNew Product - EXR125120 g0,01 mg80×80 mm0,02 mg0,2 mg5 s372×408×229 mm1 * 1 items7 180,00 €New Product
Specifications
Cat. No.OHAU31059122ModelEXR125Operating temperature range10...30 °CCommunication interfaceEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Voltage100 - 240 VNew Product - EXR225D120 /220 g0,01 mg / 0,1 mg80×80 mm0,1 mg0,2 mg5 s372×408×229 mm1 * 1 items7 710,00 €New Product
Specifications
Cat. No.OHAU31059123ModelEXR225DOperating temperature range10...30 °CCommunication interfaceEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Voltage100 - 240 VNew Product - EXR225220 g0,01 mg80×80 mm0,02 mg0,2 mg5 s372×408×229 mm1 * 1 items9 470,00 €New Product
Specifications
Cat. No.OHAU31059124ModelEXR225Operating temperature range10...30 °CCommunication interfaceEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Voltage100 - 240 VNew Product
Approved Versions
- ModelWeighing cap.ReadabilityPan/Platform SizeRepeatabilityLinearityStabilisation timeDimensions
- EXR125DM82 /120 g0,01 mg / 0,1 mg80×80 mm0,1 mg0,2 mg5 s372×408×229 mm1 * 1 items6 560,00 €New Product
Specifications
Cat. No.OHAU31059132ModelEXR125DMOperating temperature range10...30 °CCommunication interfaceEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Voltage100 - 240 VNew Product - EXR125M120 g0,01 mg80×80 mm0,02 mg0,2 mg5 s372×408×229 mm1 * 1 items7 590,00 €New Product
Specifications
Cat. No.OHAU31059133ModelEXR125MOperating temperature range10...30 °CCommunication interfaceEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Voltage100 - 240 VNew Product - EXR225DM120 /220 g0,01 mg / 0,1 mg80×80 mm0,1 mg0,2 mg5 s372×408×229 mm1 * 1 items8 190,00 €New Product
Specifications
Cat. No.OHAU31059134ModelEXR225DMOperating temperature range10...30 °CCommunication interfaceEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Voltage100 - 240 VNew Product - EXR225M220 g0,01 mg80×80 mm0,02 mg0,2 mg5 s372×408×229 mm1 * 1 items9 950,00 €New Product
Specifications
Cat. No.OHAU31059135ModelEXR225MOperating temperature range10...30 °CCommunication interfaceEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Voltage100 - 240 VNew Product