Order Entry
Export
Orders LinkContactUsLinkComponent
 

NCH 51 ≥99% HDAC inhibitor

Supplier:   Abcam
NCH 51 ≥99% HDAC inhibitor
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

Novel HDAC inhibitor (IC₅₀ values are 32, 48 and 41 nM at HDAC4, HDAC1 and HDAC6, respectively). Induces apoptosis and inhibits cell growth of various malignant cells and tumor cells lines (EC₅₀ = 3,8 μM). Also reactivates latent HIV-1 gene expression.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  PTACH, Cpd 51, S-(7-Oxo-7-((4-phenylthiazol-2-yl)amino)heptyl) 2-methylpropanethioate, 2-Methylpropanethioic acid S-[7-oxo-7-[(4-phenyl-2-thiazolyl)amino]heptyl] ester
  • CAS Number:848354-66-5
  • Molecular Weight:390,6 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% PTACH
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 185,00 €
    185,00 €
    1 * 10 mg
       185,00 €

  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 836,00 €
    836,00 €
    1 * 50 mg
       836,00 €