Order Entry
Export
Orders LinkContactUsLinkComponent
TW-37 ≥98%

TW-37 ≥98%

Supplier:   Abcam
TW-37 ≥98%
Supplier:  Abcam
Certificates

4 Options Available Below

Product Details & Documents

Small-molecule inhibitor of BCL-2 (Ki = 0,29 μM). Inhibits multiple BCL-2 family members including BCL-XL (Ki = 1,11 μM) and MCL-1 (Ki= 0,26 μM). Binds to the BCL-2 homology domain 3 (BH3) groove of BCL-2. Significant anti-proliferative and pro-apoptotic activity in lymphoma cells.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  N-[4-(2-tert-Butylphenylsulphonyl)phenyl]-2,3,4-trihydroxy-5-(2-isopropylbenzyl)benzamide
  • CAS Number:877877-35-5
  • Molecular Weight:573,7 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    98% TW-37
Product Family Options

ibitors/Activators

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 190,00 €
    190,00 €
    1 * 10 mg
       190,00 €

  • Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 436,00 €
    436,00 €
    1 * 25 mg
       436,00 €

  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 785,00 €
    785,00 €
    1 * 50 mg
       785,00 €

  • Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 1 120,00 €
    1 120,00 €
    1 * 100 mg
       1 120,00 €