Order Entry
Export
Orders LinkContactUsLinkComponent
RO 20-1724 ≥99% Phosphodiesterase 4 (PDE4) inhibitor

RO 20-1724 ≥99% Phosphodiesterase 4 (PDE4) inhibitor

Supplier:   Abcam
RO 20-1724 ≥99% Phosphodiesterase 4 (PDE4) inhibitor
Supplier:  Abcam
Certificates
RO 20-1724 ≥99% Phosphodiesterase 4 (PDE4) inhibitor

2 Options Available Below

Product Details & Documents

Selective cAMP-specific phosphodiesterase 4 (PDE4) inhibitor (IC₅₀ = 3 μM). Centrally active in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:29925-17-5
  • Molecular Weight:278,35 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% 4-(3-Butoxy-4-methoxybenzyl)imidazolidin-2-one
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 123,00 €
    123,00 €
    1 * 50 mg
       123,00 €

  • Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 467,00 €
    467,00 €
    1 * 250 mg
       467,00 €