Order Entry
Export
Orders LinkContactUsLinkComponent
 

FLUO 3/AM ≥95% calcium sensor dye

Catalog # ABCAAB145254-1
Supplier:   Abcam
FLUO 3/AM ≥95% calcium sensor dye
Catalog # ABCAAB145254-1
Supplier:  Abcam
CAS Number:  121714-22-5
FLUO 3/AM ≥95% calcium sensor dye
CAS Number:  121714-22-5
Price
467,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Cell-permeable acetoxymethyl (AM) ester of the fluorescent calcium indicator Fluo-3, Commonly used in cell-based experiments to detect changes in intracellular calcium levels. Practically nonfluorescent until it is hydrolysed intracellularly to Fluo-3 and in the presence of Ca²+.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  4-(6-Acetoxymethoxy-2,7-dichloro-3-oxo-9-xanthenyl)-4′-methyl-2,2′(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, 1-[Amino-5-(2,7-dichloro-6-acetomethoxy-3-oxo-3H-xanthen-9-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid pentaacetoxy-methyl ester
  • CAS Number:121714-22-5
  • Molecular Weight:1129,85 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Fluo-3 AM