Order Entry
Export
Orders LinkContactUsLinkComponent
 

Oligomycin ≥95% ATP synthase inhibitor

Supplier:   Abcam
Oligomycin ≥95% ATP synthase inhibitor
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

ATP synthase inhibitor. Blocks the proton channel (Fo subunit). Macrolide antibiotic.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Oligomycin A, B, C, Oligomycin complex
  • CAS Number:1404-19-9
  • Molecular Weight:791,06 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    67,5% Oligomycin A, 19,4% Oligomycin B, 9,8% Oligomycin A, 12-deoxy-
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 92,30 €
    92,30 €
    1 * 1 mg
       92,30 €

ibitors/Activators

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 164,00 €
    164,00 €
    1 * 5 mg
       164,00 €