Order Entry
Export
Orders LinkContactUsLinkComponent
 

Apoptosis activator 2 ≥99% Cytochrome c caspase activator and apoptosis inducer

Supplier:   Abcam
Apoptosis activator 2 ≥99% Cytochrome c caspase activator and apoptosis inducer
Supplier:  Abcam
Certificates
Apoptosis activator 2 ≥99% Cytochrome c caspase activator and apoptosis inducer

2 Options Available Below

Product Details & Documents

Cell-permeable cytochrome c caspase activator. Promoting cytochrome-c dependent oligomerisation of Apaf-1 into the mature apoptosome. Induces apoptosis in tumor cells (IC₅₀ = ~4 μM).

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione, 1-(3,4-Dichlorobenzyl)indoline-2,3-dione
  • CAS Number:79183-19-0
  • Molecular Weight:306,1 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Apoptosis Activator 2 (AAII)
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 164,00 €
    164,00 €
    1 * 10 mg
       164,00 €

unology General

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 728,00 €
    728,00 €
    1 * 50 mg
       728,00 €