Orders LinkContactUsLinkComponent
20-Hydroxyecdysone ≥98% Steroid invertebrate hormone
 :   Abcam
20-Hydroxyecdysone ≥98% Steroid invertebrate hormone
 :  Abcam
20-Hydroxyecdysone ≥98% Steroid invertebrate hormone
Steroid invertebrate hormone. Regulates molting, metamorphosis and reproduction in arthropods. Neuroprotective against oxidative stress in mammals. Inhibits NF-κB activation and iNOS expression. Antioxidant and apoptotic agent.
 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
 :  β-Ecdysone
  •  :5289-74-7
  •  :480,6 g/mol
  • Constituents::
    100% Ecdysterone
Product Family Options
  •  
  •    
    1 * 5 mg
    1 * 5 mg - 149,00 €
    149,00 €
    1 * 5 mg
       149,00 €
  •    
    1 * 10 mg
    1 * 10 mg - 236,00 €
    236,00 €
    1 * 10 mg
       236,00 €
  •    
    1 * 25 mg
    1 * 25 mg - 405,00 €
    405,00 €
    1 * 25 mg
       405,00 €