Order Entry
Export
Orders LinkContactUsLinkComponent
 

20-Hydroxyicosanoic acid ≥98% omega-Hydroxy C20:0 fatty acid

Catalog # ABCAAB144108-25
Supplier:   Abcam
20-Hydroxyicosanoic acid ≥98% omega-Hydroxy C20:0 fatty acid
Catalog # ABCAAB144108-25
Supplier:  Abcam
CAS Number:  62643-46-3
20-Hydroxyicosanoic acid ≥98% omega-Hydroxy C20:0 fatty acid
CAS Number:  62643-46-3
Price
405,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

ω-Hydroxy C20:0 fatty acid. For use in long chain ω-hydroxy fatty acid studies and as internal standard.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:62643-46-3
  • Molecular Weight:328,5 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% 20-hydroxyicosanoic acid