Order Entry
Export
Orders LinkContactUsLinkComponent
Xanthorrhizol ≥97% antioxidant

Xanthorrhizol ≥97% antioxidant

Catalog # ABCAAB144387-1
Supplier:   Abcam
Xanthorrhizol ≥97% antioxidant
Catalog # ABCAAB144387-1
Supplier:  Abcam
CAS Number:  30199-26-9
Xanthorrhizol ≥97% antioxidant
CAS Number:  30199-26-9
Price
272,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Antioxidant agent (IC₅₀ = 2 μM, copper-mediated oxidation of LDL). Antiproliferative agent. Activates caspase-3 and caspase-9. Modulates PARP-1 protein. Induces cytochrome c release. Shows apoptotic, antibacterial, antifungal, anticancer, phytoestrogenic, neuroprotective, antihyperglycemic and anti-infla mMatory effects in vivo. Orally active.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:30199-26-9
  • Molecular Weight:218,33 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% 2-methyl-5-[(2R)-6-methylhept-5-en-2-yl]phenol