Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Pyrazoloanthrone ≥99% JNK inhibitor

1,9-Pyrazoloanthrone ≥99% JNK inhibitor

Supplier:   Abcam
1,9-Pyrazoloanthrone ≥99% JNK inhibitor
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

Potent and selective JNK1, -2, and -3 inhibitor (IC₅₀ = 0,11 μM). SP600125 is a reversible ATP-competitive inhibitor with 20-fold selectivity over a range of kinases. It dose-dependently inhibits the phosphorylation of c-Jun and the expression of infla mMatory genes COX-2, IL-2, IFNγ and TNFα. Neuroprotective in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125
  • CAS Number:129-56-6
  • Molecular Weight:220,23 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Anthra[1,9-cd]pyrazol-6(2H)-one
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 118,00 €
    118,00 €
    1 * 10 mg
       118,00 €

  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 446,00 €
    446,00 €
    1 * 50 mg
       446,00 €