Amphotericin B ≥90% Antimycotic polyene
About this item
Antimycotic polyene. Disrupts fungal cell membrane permeability by targeting sterols such as ergosterol. Active in vivo.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:1397-89-3
- Molecular Weight:924,1 g/mol
2 Options Available Below
Product Details & Documents
Antimycotic polyene. Disrupts fungal cell membrane permeability by targeting sterols such as ergosterol. Active in vivo.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:1397-89-3
- Molecular Weight:924,1 g/mol
Specifications for Product Family
Specification Test Results
- Constituents::100% Amphotericin B
Product Family Options
Product Information
- AvailabilityPrice



