Order Entry
Export
Orders LinkContactUsLinkComponent
(-)-Quinine ≥98% Potassium channel blocker

(-)-Quinine ≥98% Potassium channel blocker

Catalog # ABCAAB141247-5
Supplier:   Abcam
(-)-Quinine ≥98% Potassium channel blocker
Catalog # ABCAAB141247-5
Supplier:  Abcam
CAS Number:  130-95-0
(-)-Quinine ≥98% Potassium channel blocker
CAS Number:  130-95-0
Price
134,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Potassium channel blocker. Blocks small conductance Ca²⁺ -activated K⁺ channels. Inhibits TRPM7 currents. Shows antipyretic, antimalarial, analgesic, anti-infla mMatory and anticonvulsant effects in vivo. Orally active.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol
  • CAS Number:130-95-0
  • Molecular Weight:324,42 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Quinine