Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl blue stain for histology

Methyl blue stain for histology

Catalog # ABCAAB146271-50
Supplier:   Abcam
Methyl blue stain for histology
Catalog # ABCAAB146271-50
Supplier:  Abcam
CAS Number:  28983-56-4
Methyl blue stain for histology
CAS Number:  28983-56-4
Price
76,90 €
Log in for availability
1 * 50 g
Product Details & Documents

Histology stain for collagen in tissue sections. Used in some differential staining techniques such as Mallory′s connective tissue stain and Gomori′s one-step trichrome procedure, and has been incorporated in Sabouraud agar for the identification of Candida albicans strains.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Aniline blue sodium salt, [[4-[bis[4-[(sulphophenyl)amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]amino]-benzenesulphonic acid sodium salt (1:2), Poirriers blue, Cotton blue, Water blue, C.I. 42780, Aniline blue, water soluble, Disodium [[4-[bis[4-[(sulphonatophenyl)amino]phenyl]methylene]cyclohexa-2,5-dien-1-ylidene]amino]benzenesulphonate, Acid Blue 93
  • CAS Number:28983-56-4
  • Molecular Weight:799,8 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Methyl blue sodium salt