Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Aminobenzhydrazide ≥95% MPO inhibitor

Supplier:   Abcam
4-Aminobenzhydrazide ≥95% MPO inhibitor
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

Potent, irreversible and specific MPO inhibitor (IC₅₀ = 300 nM). Benzoic acid analog. No inhibition of catalase or glutathione peroxidase activity. Inhibits apoptosis in vitro.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  4-Aminobenzoic hydrazide, 4-Aminobenzene-1-carbohydrazide, 4-Aminobenzoic acid hydrazide, 4-Aminobenzohydrazide
  • CAS Number:5351-17-7
  • Molecular Weight:151,17 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% 4-Aminobenzoic hydrazide
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 123,00 €
    123,00 €
    1 * 100 mg
       123,00 €

  • Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 334,00 €
    334,00 €
    1 * 500 mg
       334,00 €