Order Entry
Export
Orders LinkContactUsLinkComponent
 

Linoleic acid ≥99% PPAR activator

Catalog # ABCAAB141144-1
Supplier:   Abcam
Linoleic acid ≥99% PPAR activator
Catalog # ABCAAB141144-1
Supplier:  Abcam
CAS Number:  60-33-3
Linoleic acid ≥99% PPAR activator
CAS Number:  60-33-3
Price
113,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Essential polyunsaturated fatty acid. PPAR activator. Induces apoptosis. Augments cellular oxidant status and induces mitochondrial dysfunction. Modulates the infla mMatory process and regulates neutrophil function. Stimulates thromboxanes and prostaglandin production. Shows antithrombotic effects in vivo. Orally active.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
  • CAS Number:60-33-3
  • Molecular Weight:280,4 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Linoleic acid