Order Entry
Export
Orders LinkContactUsLinkComponent
 

L(-)-Tryptophan ≥99% essential amino acid

Catalog # ABCAAB146400-50
Supplier:   Abcam
L(-)-Tryptophan ≥99% essential amino acid
Catalog # ABCAAB146400-50
Supplier:  Abcam
CAS Number:  73-22-3
L(-)-Tryptophan ≥99% essential amino acid
CAS Number:  73-22-3
Price
76,90 €
Log in for availability
1 * 50 g
Product Details & Documents

Essential amino acid which is necessary for normal growth in infants and for nitrogen balance in adults. Used as an antidepressant, anxiolytic and sleep aid. Precursor to niacin, indole alkaloids, serotonin and melatonin.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
  • CAS Number:73-22-3
  • Molecular Weight:204,2 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% L-Tryptophan