Order Entry
Export
Orders LinkContactUsLinkComponent
L(+)-Glutamic acid ≥99% excitatory neurotransmitter

L(+)-Glutamic acid ≥99% excitatory neurotransmitter

Supplier:   Abcam
L(+)-Glutamic acid ≥99% excitatory neurotransmitter
Supplier:  Abcam
Certificates
L(+)-Glutamic acid ≥99% excitatory neurotransmitter

2 Options Available Below

Product Details & Documents

Major excitatory neurotransmitter. Acts at glutamate receptors. Active in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
  • CAS Number:56-86-0
  • Molecular Weight:147,13 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% L-Glutamate
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 2.500 mg
    1 * 2.500 mg - 144,00 €
    144,00 €
    1 * 2.500 mg
       144,00 €

  • Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 66,70 €
    66,70 €
    1 * 1 g
       66,70 €